missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid, 97%, Maybridge

Manufacturer:  Thermo Scientific CC29801DA

 View more versions of this product

Catalog No. CC29801DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-2-carboxylic acid
1g
96.5
97%
MFCD11506351
SSVIRLKDUUXSTR-UHFFFAOYSA-N
4-methyl-2,3-dihydro-1,4-benzoxazine-2-carboxylic acid
10845449
97%
Amber Glass Bottle
212578-38-6
100.0
C10H11NO3
4-methyl-3,4-dihydro-2h-1,4-benzoxazine-2-carboxylic acid, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine-2-carboxylic acid, 2h-1,4-benzoxazine-2-carboxylic acid, 3,4-dihydro-4-methyl, 4-methyl-2h,3h-benzo e 1,4-oxazine-2-carboxylic acid, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-2-carboxylic aci, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-2-carboxylic acid, 3,4-dihydro-4-methyl-2h-1,4-benzoxazole-2-carboxylic acid
CN1CC(OC2=CC=CC=C21)C(=O)O
193.202
193.2

Chemical Identifiers

212578-38-6
193.202
SSVIRLKDUUXSTR-UHFFFAOYSA-N
10845449
CN1CC(OC2=CC=CC=C21)C(=O)O
C10H11NO3
MFCD11506351
4-methyl-3,4-dihydro-2h-1,4-benzoxazine-2-carboxylic acid, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine-2-carboxylic acid, 2h-1,4-benzoxazine-2-carboxylic acid, 3,4-dihydro-4-methyl, 4-methyl-2h,3h-benzo e 1,4-oxazine-2-carboxylic acid, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-2-carboxylic aci, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-2-carboxylic acid, 3,4-dihydro-4-methyl-2h-1,4-benzoxazole-2-carboxylic acid
4-methyl-2,3-dihydro-1,4-benzoxazine-2-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents