missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-2-thienylboronic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 162607-15-0
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C5H7BO2S
Formula Weight 141.99
Synonym 4-Methyl-2-thienylboric acid
Chemical Name or Material 4-Methylthiophene-2-boronic acid
Assay ≥95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC359605000 SDS Thermo Fisher Scientific
35960-5000
Glass bottle 500mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

162607-15-0
100.0
C5H7BO2S
4-Methyl-2-thienylboric acid
≥95%
95.0
96%
141.99
4-Methylthiophene-2-boronic acid

Specifications

141.99
500mg
100.0
Authentic
162607-15-0
4-Methyl-2-thienylboric acid
4-Methylthiophene-2-boronic acid
Powder
White to light beige
95.0
C5H7BO2S
Glass bottle
96%
≥95%