Please login to your online account to display your discounted pricing

(4-Methyl-2-pyrid-4-yl-1,3-thiazol-5-yl)methylamine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material (4-Methyl-2-pyrid-4-yl-1, 3-thiazol-5-yl)methylamine
CAS 933742-59-7
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H11N3S
Formula Weight 205.28
MDL Number MFCD09879909
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC58613CB

  SDS
fisher scientific company
CC58613CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC58613DA

  SDS
fisher scientific company
CC58613DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 205.28
Quantity 250mg
Assay 97%
CAS Max % 100.0
Chemical Name or Material (4-Methyl-2-pyrid-4-yl-1, 3-thiazol-5-yl)methylamine
Assay Percent Range 97%
Molecular Formula C10H11N3S
CAS 933742-59-7
Packaging Amber Glass Bottle
MDL Number MFCD09879909
CAS Min % 96.5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.