missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(4-Methyl-2-pyrid-4-yl-1,3-thiazol-5-yl)methanol, 97%, Maybridge

Chemical Identifiers

CAS 886851-57-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H10N2OS
Formula Weight 206.27
MDL Number MFCD09817497
Chemical Name or Material (4-Methyl-2-pyrid-4-yl-1, 3-thiazol-5-yl)methanol
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC58609CB SDS Fisher Scientific Company
CC58609CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC58609DA SDS Fisher Scientific Company
CC58609DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

886851-57-6
100.0
C10H10N2OS
MFCD09817497
97%
96.5
97%
206.27
(4-Methyl-2-pyrid-4-yl-1, 3-thiazol-5-yl)methanol

Specifications

206.27
100.0
C10H10N2OS
Amber Glass Bottle
97%
97%
250mg
886851-57-6
96.5
MFCD09817497
(4-Methyl-2-pyrid-4-yl-1, 3-thiazol-5-yl)methanol

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning