missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-2-pyrid-3-yl-1,3-thiazole-5-carbaldehyde, 97%, Maybridge™

Manufacturer:  Fisher Scientific Company CC74104DA

 View more versions of this product

Catalog No. CC74104DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Methyl-2-pyrid-3-yl-1, 3-thiazole-5-carbaldehyde
1g
96.5
97%
MFCD12198119
KHHZRQVJLBGSNM-UHFFFAOYSA-N
4-methyl-2-pyridin-3-yl-1,3-thiazole-5-carbaldehyde
43811058
97%
Amber Glass Bottle
958443-39-5
100.0
C10H8N2OS
4-methyl-2-3-pyridyl-1,3-thiazole-5-carbaldehyde, 4-methyl-2-pyrid-3-yl-1,3-thiazole-5-carbaldehyde, 4-methyl-2-pyridin-3-yl thiazole-5-carbaldehyde, 4-methyl-2-pyridin-3-yl-1,3-thiazole-5-carbaldehyde, 4-methyl-2-pyridin-3-yl-thiazole-5-carbaldehyde
CC1=C(SC(=N1)C2=CN=CC=C2)C=O
204.247
204.25

Chemical Identifiers

958443-39-5
204.247
KHHZRQVJLBGSNM-UHFFFAOYSA-N
43811058
CC1=C(SC(=N1)C2=CN=CC=C2)C=O
C10H8N2OS
MFCD12198119
4-methyl-2-3-pyridyl-1,3-thiazole-5-carbaldehyde, 4-methyl-2-pyrid-3-yl-1,3-thiazole-5-carbaldehyde, 4-methyl-2-pyridin-3-yl thiazole-5-carbaldehyde, 4-methyl-2-pyridin-3-yl-1,3-thiazole-5-carbaldehyde, 4-methyl-2-pyridin-3-yl-thiazole-5-carbaldehyde
4-methyl-2-pyridin-3-yl-1,3-thiazole-5-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents