Please login to your online account to display your discounted pricing

(4-methyl-2-pyrid-3-yl-1,3-thiazol-5-yl)methylamine, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC74113DE

 View more versions of this product

Catalog No. CC74113DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 205.28
Quantity 5g
Assay 97%
Molecular Formula C10H11N3S
Packaging Amber Glass Bottle
Chemical Name or Material (4-methyl-2-pyrid-3-yl-1, 3-thiazol-5-yl)methylamine
CAS 108214124
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material (4-methyl-2-pyrid-3-yl-1, 3-thiazol-5-yl)methylamine
Assay 97%
Molecular Formula C10H11N3S
Formula Weight 205.28
Accessories

GHS Signal Word: Danger

SDS & Safety Documents