Please login to your online account to display your discounted pricing

(4-methyl-2-pyrid-3-yl-1,3-thiazol-5-yl)methylamine, Maybridge

Chemical Identifiers

Chemical Name or Material (4-methyl-2-pyrid-3-yl-1, 3-thiazol-5-yl)methylamine
Assay 97%
Molecular Formula C10H11N3S
Formula Weight 205.28
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC74113DA

  SDS
fisher scientific company
CC74113DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC74113CB

  SDS
fisher scientific company
CC74113CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC74113DE

  SDS
fisher scientific company
CC74113DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 205.28
Quantity 1g
Assay 97%
Molecular Formula C10H11N3S
Chemical Name or Material (4-methyl-2-pyrid-3-yl-1, 3-thiazol-5-yl)methylamine
Packaging Amber Glass Bottle
CAS 108214124
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger