Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-2-phenylthiazole, 98%

$56.50 - $200.41

Chemical Identifiers

Chemical Name or Material 4-Methyl-2-phenylthiazole
CAS 1826-17-1
Molecular Formula C10H9NS
Formula Weight 175.25
MDL Number MFCD00272328
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1957206

  SDS
alfa aesar™
A1957206
5g Each for $200.41

AAA1957203

  SDS
alfa aesar™
A1957203
1g Each for $56.50
Description & Specifications

Specifications

Formula Weight 175.25
MDL Number MFCD00272328
Quantity 5g
Assay 98%
Chemical Name or Material 4-Methyl-2-phenylthiazole
Molecular Formula C10H9NS
CAS 1826-17-1