Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-2-phenylimidazole, 95%


Quantity 10g

Manufacturer: alfa aesar™  L1294509

 View more versions of this product

Catalog No. AAL1294509

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 158.2
Chemical Name or Material 4-Methyl-2-phenylimidazole
TSCA TSCA
Assay 95%
Molecular Formula C10H10N2
MDL Number MFCD00047015
CAS 827-43-0
Beilstein 4112
Health Hazard 1 Warning
Melting Point 174° to 176°C
Quantity 10g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material 4-Methyl-2-phenylimidazole
CAS 827-43-0
Molecular Formula C10H10N2
Formula Weight 158.2
MDL Number MFCD00047015
Beilstein 4112
Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents