Please login to your online account to display your discounted pricing

(4-Methyl-2-phenyl-5-pyrimidinyl)methanol, 95%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  CC20209DE

 View more versions of this product

Catalog No. CC20209DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 200.24
Quantity 5g
Assay 95%
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C12H12N2O
CAS Min % 94.5
MDL Number MFCD02682074
Chemical Name or Material (4-Methyl-2-phenyl-5-pyrimidinyl)methanol
CAS 342405-27-0

Specifications

Chemical Name or Material (4-Methyl-2-phenyl-5-pyrimidinyl)methanol
CAS 342405-27-0
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C12H12N2O
Formula Weight 200.24
MDL Number MFCD02682074