Please login to your online account to display your discounted pricing

4-Methyl-2-phenyl-5-pyrimidinecarboxylic acid, 97%


Quantity 5g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC20201DE

 View more versions of this product

Catalog No. CC20201DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 214.22
Quantity 5g
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H10N2O2
Packaging Amber glass bottle
Chemical Name or Material 4-Methyl-2-phenyl-5-pyrimidinecarboxylic acid
CAS 103249-79-2

Specifications

Chemical Name or Material 4-Methyl-2-phenyl-5-pyrimidinecarboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H10N2O2
Formula Weight 214.22