missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-2-phenyl-5-pyrimidinecarboxylic acid, 97%

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C12H10N2O2
Formula Weight 214.22
Chemical Name or Material 4-Methyl-2-phenyl-5-pyrimidinecarboxylic acid
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC20201DA SDS Fisher Scientific Company
CC20201DA
Amber glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC20201DE SDS Fisher Scientific Company
CC20201DE
Amber glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC20201EA Fisher Scientific Company
CC20201EA
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
214.22
97%
C12H10N2O2
4-Methyl-2-phenyl-5-pyrimidinecarboxylic acid

Specifications

1g
Amber glass bottle
C12H10N2O2
4-Methyl-2-phenyl-5-pyrimidinecarboxylic acid
103249-79-2
97%
214.22
97%