Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-2-phenyl-2-pentenal, (E)+(Z), 90+%

$59.51 - $191.36

Chemical Identifiers

Chemical Name or Material 4-Methyl-2-phenyl-2-pentenal
CAS 26643-91-4
Name Note (E)+(Z)
Molecular Formula C12H14O
Formula Weight 174.24
MDL Number MFCD00051834
Beilstein 2042583
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1873809

  SDS
alfa aesar™
A1873809
10g Each for $59.51

AAA1873818

  SDS
alfa aesar™
A1873818
50g Each for $191.36
Description & Specifications

Specifications

Formula Weight 174.24
TSCA TSCA
Molecular Formula C12H14O
Name Note (E)+(Z)
Flash Point 110°C (230°F)
Assay ≥90%
Boiling Point 84° to 86°C (0.7mmHg)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-Methyl-2-phenyl-2-pentenal
MDL Number MFCD00051834
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 26643-91-4
Recommended Storage 2° to 8°C
Health Hazard 1 Warning
Beilstein 2042583
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C