missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methyl-2-phenyl-1,3-thiazole-5-sulfonyl chloride, Technical, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC29203DA
Specifications
4-Methyl-2-phenyl-1, 3-thiazole-5-sulfonyl chloride | |
95% | |
C10H8ClNO2S2 | |
4-methyl-2-phenylthiazole-5-sulfonyl chloride, 4-methyl-2-phenyl-thiazole-5-sulfonyl chloride, 5-thiazolesulfonyl chloride, 4-methyl-2-phenyl, 4-methyl-2-phenyl-1,3-thiazole-5-sulfonylchloride | |
CC1=C(SC(=N1)C2=CC=CC=C2)S(=O)(=O)Cl | |
273.749 | |
273.76 |
690632-88-3 | |
Amber Glass Bottle | |
1g | |
NGDQQLAVJWUYSF-UHFFFAOYSA-N | |
4-methyl-2-phenyl-1,3-thiazole-5-sulfonyl chloride | |
2795490 | |
95% |
Chemical Identifiers
690632-88-3 | |
273.749 | |
4-methyl-2-phenylthiazole-5-sulfonyl chloride, 4-methyl-2-phenyl-thiazole-5-sulfonyl chloride, 5-thiazolesulfonyl chloride, 4-methyl-2-phenyl, 4-methyl-2-phenyl-1,3-thiazole-5-sulfonylchloride | |
4-methyl-2-phenyl-1,3-thiazole-5-sulfonyl chloride |
C10H8ClNO2S2 | |
NGDQQLAVJWUYSF-UHFFFAOYSA-N | |
2795490 | |
CC1=C(SC(=N1)C2=CC=CC=C2)S(=O)(=O)Cl |