Please login to your online account to display your discounted pricing

4-Methyl-2-phenyl-1,3-thiazole-5-carbaldehyde, Maybridge

Chemical Identifiers

Chemical Name or Material 4-Methyl-2-phenyl-1, 3-thiazole-5-carbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H9NOS
Formula Weight 203.26
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC29204DA

  SDS
fisher scientific company
CC29204DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC29204CB

  SDS
fisher scientific company
CC29204CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.26
Quantity 1g
Assay 97%
Chemical Name or Material 4-Methyl-2-phenyl-1, 3-thiazole-5-carbaldehyde
Molecular Formula C11H9NOS
CAS 55327-23-6
Assay Percent Range 97%
Packaging Amber Glass Bottle