missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-2-phenyl-1,3-oxazole-5-carbonyl chloride, 97%, Maybridge

Chemical Identifiers

CAS 52169-89-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H8ClNO2
Formula Weight 221.64
MDL Number MFCD10700047
Chemical Name or Material 4-Methyl-2-phenyl-1, 3-oxazole-5-carbonyl chloride
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC18102DA Fisher Scientific Company
CC18102DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

52169-89-8
100.0
C11H8ClNO2
MFCD10700047
97%
96.5
97%
221.64
4-Methyl-2-phenyl-1, 3-oxazole-5-carbonyl chloride

Specifications

1g
96.5
Amber Glass Bottle
C11H8ClNO2
MFCD10700047
97%
52169-89-8
100.0
97%
221.64
4-Methyl-2-phenyl-1, 3-oxazole-5-carbonyl chloride

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger