Please login to your online account to display your discounted pricing

4-Methyl-2-phenyl-1,3-oxazole-5-carbaldehyde, 95%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC18104CB

 View more versions of this product

Catalog No. CC18104CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 187.2
Quantity 250mg
Assay 95%
CAS Max % 100.0
Chemical Name or Material 4-Methyl-2-phenyl-1, 3-oxazole-5-carbaldehyde
Assay Percent Range 95%
Molecular Formula C11H9NO2
MDL Number MFCD11109318
CAS 953408-85-0
CAS Min % 94.5
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 4-Methyl-2-phenyl-1, 3-oxazole-5-carbaldehyde
CAS 953408-85-0
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H9NO2
Formula Weight 187.2
MDL Number MFCD11109318