missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(4-Methyl-2-phenyl-1,3-oxazol-5-ylmethyl)amine, 97%, Maybridge

Chemical Identifiers

CAS 165735-97-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H12N2O
Formula Weight 188.23
MDL Number MFCD11109320
Chemical Name or Material (4-Methyl-2-phenyl-1, 3-oxazol-5-ylmethyl)amine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC18113DA Fisher Scientific Company
CC18113DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC18113CB Fisher Scientific Company
CC18113CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

165735-97-7
100.0
C11H12N2O
MFCD11109320
97%
96.5
97%
188.23
(4-Methyl-2-phenyl-1, 3-oxazol-5-ylmethyl)amine

Specifications

188.23
100.0
97%
MFCD11109320
C11H12N2O
97%
1g
Amber Glass Bottle
96.5
165735-97-7
(4-Methyl-2-phenyl-1, 3-oxazol-5-ylmethyl)amine

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger