Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-2-phenyl-1,2,3-triazole-5-carboxylic acid, 98%

$103.76 - $415.61

Chemical Identifiers

Chemical Name or Material 4-Methyl-2-phenyl-1,2,3-triazole-5-carboxylic acid
CAS 22300-56-7
Molecular Formula C10H9N3O2
Formula Weight 203.2
MDL Number MFCD00051653
Beilstein 177649
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0805106

  SDS
alfa aesar™
L0805106
5g Each for $415.61

AAL0805103

  SDS
alfa aesar™
L0805103
1g Each for $103.76
Description & Specifications

Specifications

Formula Weight 203.2
Chemical Name or Material 4-Methyl-2-phenyl-1,2,3-triazole-5-carboxylic acid
Assay 98%
MDL Number MFCD00051653
CAS 22300-56-7
Health Hazard 1 Warning
Molecular Formula C10H9N3O2
Beilstein 177649
Melting Point 200° to 205°C
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.