missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-2-pentyne, 97%

Manufacturer:  Alfa Aesar™ H5344214

 View more versions of this product

Catalog No. AAH5344214



Specifications

Specifications

4-Methyl-2-pentyne
−12°C (−10°F)
71°C to 73°C
21020-27-9
(CH3)2CHC≡CCH3
UN3295
SLMFWJQZLPEDDU-UHFFFAOYSA-N
4-methylpent-2-yne
140789
97%
0.711
1.407
25g
C6H10
MFCD00041615
2-pentyne, 4-methyl
CC#CC(C)C
82.146
82.15

Chemical Identifiers

21020-27-9
82.146
SLMFWJQZLPEDDU-UHFFFAOYSA-N
140789
CC#CC(C)C
C6H10
MFCD00041615
2-pentyne, 4-methyl
4-methylpent-2-yne
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents