missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-2-pentyne, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5344206

 View more versions of this product

Catalog No. AAH5344206



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Methyl-2-pentyne
0.711
−12°C (−10°F)
1.407
MFCD00041615
UN3295
SLMFWJQZLPEDDU-UHFFFAOYSA-N
4-methylpent-2-yne
140789
97%
21020-27-9
71°C to 73°C
C6H10
(CH3)2CHC≡CCH3
5 g
4-methyl-2-pentyne, 2-pentyne, 4-methyl, isopropylmethylacetylene, acmc-1co71
CC#CC(C)C
82.15
82.15

Chemical Identifiers

21020-27-9
82.15
SLMFWJQZLPEDDU-UHFFFAOYSA-N
140789
CC#CC(C)C
C6H10
MFCD00041615
4-methyl-2-pentyne, 2-pentyne, 4-methyl, isopropylmethylacetylene, acmc-1co71
4-methylpent-2-yne
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only