missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-2-pentene (cis- and trans- mixture), TCI America™

Manufacturer:  TCI America M03955ML

 View more versions of this product

Catalog No. M03955ML



Specifications

Specifications

4-Methyl-2-pentene (cis- and trans- mixture)
Colorless
4461-48-7
MFCD00065138
LGAQJENWWYGFSN-PLNGDYQASA-N
(Z)-4-methylpent-2-ene
5326159
Clear Liquid at 20°C
58°C
5mL
C6H12
2288
CC=CC(C)C
84.162
84.16
≥96.0% (GC)

Chemical Identifiers

4461-48-7
84.162
LGAQJENWWYGFSN-PLNGDYQASA-N
(Z)-4-methylpent-2-ene
C6H12
MFCD00065138
5326159
CC=CC(C)C
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0031

TSCA : No

Documents