missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-2-pentanone, TCI America™

$41.14 - $41.14

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00008938
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym 2-methyl-4-pentanone
PubChem CID 7909
ChEBI CHEBI:82344
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(=O)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M0389500ML View Documents Tci America
M0389500ML
500mL Each for $41.14
 
Specifications

Chemical Identifiers

108-10-1
100.161
NTIZESTWPVYFNL-UHFFFAOYSA-N
7909
4-methylpentan-2-one
C6H12O
MFCD00008938
2-methyl-4-pentanone
CHEBI:82344
CC(C)CC(=O)C

Specifications

116°C
-85°C
108-10-1
MFCD00008938
2-methyl-4-pentanone
CC(C)CC(=O)C
100.161
CHEBI:82344
Clear Liquid at 20°C
4-Methyl-2-pentanone
Colorless
500mL
C6H12O
1245
NTIZESTWPVYFNL-UHFFFAOYSA-N
4-methylpentan-2-one
7909
100.16
≥99.5% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0542

RTECSNumber : SA9275000

TSCA : Yes