missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-2-pentanone, HPLC Grade, >99%, Alfa Aesar™

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.1%, Color (APHA) 10

Chemical Identifiers

CAS 108-10-1
Physical Form Liquid
Assay Percent Range >99%
Molecular Formula C6H12O
Formula Weight 100.16
MDL Number MFCD00008938
Chemical Name or Material 4-Methyl-2-pentanone, Filtered through 0.2μ filters.
Grade HPLC Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA43170K2 SDS Alfa Aesar
43170K2
1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA43170K7 SDS Alfa Aesar
43170K7
4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA43170M1 SDS Alfa Aesar
43170M1
4 x 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA43170M6 SDS Alfa Aesar
43170M6
4 x 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

108-10-1
>99%
100.16
4-Methyl-2-pentanone, Filtered through 0.2μ filters.
Liquid
C6H12O
MFCD00008938
HPLC Grade

Specifications

Colorless liquid
Clear colorless
108-10-1
Mild
Liquid
C6H12O
MFCD00008938
4-Methyl-2-pentanone, Filtered through 0.2μ filters.
0.801g/cm3 at 20°C
1L
460°C (860°F)
21 hPa (16mm Hg) at 20°C
>99%
100.16
4-Methyl-2-pentanone
HPLC Grade

Safety and Handling

H225-H332-H319-H335: Highly flammable liquid and vapour. Harmful if inhaled. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 203-550-1