Learn More
4-Methyl-2-pentanone, HPLC Grade, 99+%, Thermo Scientific Chemicals
Maximum level of impurities: Evaporation residue 5ppm, H2O 0.1%, Color (APHA) 10
Supplier: Thermo Scientific Chemicals 043170K7
Description
4-Methyl-2-pentanone is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Methyl-2-pentanone | |
108-10-1 | |
Colorless | |
117°C to 118°C | |
460°C (860°F) | |
Mild | |
1.396 | |
4 L | |
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon | |
NTIZESTWPVYFNL-UHFFFAOYSA-N | |
4-methylpentan-2-one | |
7909 | |
100.16 | |
HPLC | |
Liquid |
Filtered through 0.2μ filters. | |
°C to 84°C | |
0.801 | |
13°C (55°F) | |
>99% | |
C6H12O | |
MFCD00008938 | |
605399 | |
Miscible with ethanol,ether,acetone,benzene and chloroform. Slightly miscible with water. | |
CC(C)CC(=O)C | |
100.161 | |
CHEBI:82344 | |
21 hPa (16mm Hg) at 20°C | |
4-Methyl-2-pentanone |
Chemical Identifiers
108-10-1 | |
100.161 | |
NTIZESTWPVYFNL-UHFFFAOYSA-N | |
7909 | |
4-methylpentan-2-one |
C6H12O | |
MFCD00008938 | |
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon | |
CHEBI:82344 | |
CC(C)CC(=O)C |
Safety and Handling
P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P370+P378q-P501c
H225-H319-H332-H335-H351
DOTInformation : DOT Class: 3, Packaging Group: II
EINECSNumber : 203-550-1
RTECSNumber : SA9275000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only