missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Riedel-de Haen™ 4-Methyl-2-pentanone, ACS Reagent, ≥98.5%, Honeywell™ Riedel-de Haën™

ACS Reagent,≥98.5%

$866.59 - $866.59

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00008938
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
PubChem CID 7909
ChEBI CHEBI:82344
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(=O)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60046616 View Documents Honeywell Riedel-de Haen™
36051120L
20L Stainless Steel Can Each for $866.59
 
Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Chemical Identifiers

108-10-1
100.161
NTIZESTWPVYFNL-UHFFFAOYSA-N
7909
4-methylpentan-2-one
C6H12O
MFCD00008938
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
CHEBI:82344
CC(C)CC(=O)C

Specifications

0.801g/mL at 25°C
Conforms to structure
n20/D 1.395
Undesignated
20L
C6H12O
MFCD00008938
605399
NTIZESTWPVYFNL-UHFFFAOYSA-N
4-methylpentan-2-one
7909
100.16g/mol
4-Methyl-2-pentanone
14°C (57.2°F)
Stainless Steel Can
118°C
-80°C
108-10-1
(CH3)2CHCH2COCH3
UN1245
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
CC(C)CC(=O)C
100.161
CHEBI:82344
≥98.5%
Safety and Handling

Safety and Handling

P210-P261-P305 + P351 + P338

DOTInformation : class 3 - PG 2 - Methyl isobutyl ketone

RTECSNumber : 203-550-1