Please login to your online account to display your discounted pricing

4-Methyl-2-pentanone, 99+%, electronic use grade, residue free, ACROS Organics™

$148.67 - $148.67

Chemical Identifiers

Chemical Name or Material 4-Methyl-2-pentanone
CAS 108-10-1
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99%
Assay Percent Range 99+%
Molecular Formula C6H12O
Linear Formula (CH3)2CHCH2COCH3
Formula Weight 100.16
MDL Number MFCD00008938
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC327920010

  SDS
thermo fisher scientific
327920010
1L Each for $148.67

AC327920025

  SDS
thermo fisher scientific
AC327920025
2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99%
Physical Form Liquid
Formula Weight 100.16
Quantity 1L
Color Clear
Beilstein 01, 691
Merck Index 15, 5253
EINECS Number 203-550-1
TSCA TSCA
Density 0.8000g/mL
RTECS Number SA9275000
CAS Min % 99.0
CAS Max % 100.0
Acidity 0.003% max.
Copper (Cu) 10ppb max.
Iron (Fe) 50ppb max.
Heavy Metals (as Pb) 1ppm max.
Assay Percent Range 99+%
Residue after Evaporation 0.001% max.
Water 0.05% max. (Coulometric)
Synonym Isobutyl methyl ketone
Arsenic (As) 10ppb max.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Barium (Ba) 20ppb max.
Chlorine (Cl) 2ppm max.
Chemical Name or Material 4-Methyl-2-pentanone
Molecular Formula C6H12O
Linear Formula (CH3)2CHCH2COCH3
MDL Number MFCD00008938
CAS 108-10-1
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Danger

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.