Please login to your online account to display your discounted pricing

4-Methyl-2-oxopentanoic acid, sodium salt, hydrate, +98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  189710100

 View more versions of this product

Catalog No. AC189710100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 152.13
Quantity 10g
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material 4-Methyl-2-oxopentanoic acid, sodium salt, hydrate
Packaging Glass bottle
Assay Percent Range 98+%
Molecular Formula C6H9NaO3·xH2O
MDL Number MFCD00150776
CAS 332360-07-3
CAS 4502-00-5
Name Note 98+%
Linear Formula (CH3)2CHCH2COCO2Na·xH2O

Specifications

Chemical Name or Material 4-Methyl-2-oxopentanoic acid, sodium salt, hydrate
Name Note 98+%
CAS 332360-07-3
CAS Min % 98.0
CAS Max % 100.0
CAS 4502-00-5
Assay Percent Range 98+%
Molecular Formula C6H9NaO3·xH2O
Linear Formula (CH3)2CHCH2COCO2Na·xH2O
Formula Weight 152.13
MDL Number MFCD00150776