missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-2-oxopentanoic acid, sodium salt, hydrate, +98%, ACROS Organics™

Chemical Identifiers

CAS 332360-07-3,4502-00-5
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Molecular Formula C6H9NaO3·xH2O
Linear Formula (CH3)2CHCH2COCO2Na·xH2O
Formula Weight 152.13
MDL Number MFCD00150776
Chemical Name or Material 4-Methyl-2-oxopentanoic acid, sodium salt, hydrate, 98+%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC189710010 SDS Fisher Scientific Acros
189710010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC189710100 SDS Thermo Fisher Scientific
189710100
Glass bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

332360-07-3,4502-00-5
100.0
C6H9NaO3·xH2O
152.13
4-Methyl-2-oxopentanoic acid, sodium salt, hydrate, 98+%
98.0
98+%
(CH3)2CHCH2COCO2Na·xH2O
MFCD00150776

Specifications

1g
98.0
Glass bottle
C6H9NaO3·xH2O
152.13
4-Methyl-2-oxopentanoic acid, sodium salt, hydrate, 98+%
332360-07-3,4502-00-5
100.0
98+%
(CH3)2CHCH2COCO2Na·xH2O
MFCD00150776