missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-2-nitrophenyl isothiocyanate, 95%

Manufacturer:  Alfa Aesar™ L1043806

 View more versions of this product

Catalog No. AAL1043806



Specifications

Specifications

4-Methyl-2-nitrophenyl isothiocyanate
62°C to 67°C
17614-74-3
MFCD00041248
2969500
CUQUTWSUXWDJKF-UHFFFAOYSA-N
1-isothiocyanato-4-methyl-2-nitrobenzene
140258
95%
Irritating
5g
C8H6N2O2S
UN2923
4-methyl-2-nitrophenyl isothiocyanate, 4-methyl-2-nitrophenylisothiocyanate, benzene,1-isothiocyanato-4-methyl-2-nitro, 4-methyl-2-nitrobenzenisothiocyanate, acmc-1brfl, 4-methy-2-nitrophenyl isothiocyanate, 1-isothiocyanato-4-methyl-2-nitro-benzene, #
CC1=CC(=C(C=C1)N=C=S)[N+](=O)[O-]
194.208
194.21

Chemical Identifiers

17614-74-3
194.208
CUQUTWSUXWDJKF-UHFFFAOYSA-N
140258
CC1=CC(=C(C=C1)N=C=S)[N+](=O)[O-]
C8H6N2O2S
MFCD00041248
4-methyl-2-nitrophenyl isothiocyanate, 4-methyl-2-nitrophenylisothiocyanate, benzene,1-isothiocyanato-4-methyl-2-nitro, 4-methyl-2-nitrobenzenisothiocyanate, acmc-1brfl, 4-methy-2-nitrophenyl isothiocyanate, 1-isothiocyanato-4-methyl-2-nitro-benzene, #
1-isothiocyanato-4-methyl-2-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

Documents