missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-2-nitroaniline, TCI America™

$42.78 - $314.73

Chemical Identifiers

CAS 89-62-3
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00007907
InChI Key DLURHXYXQYMPLT-UHFFFAOYSA-N
Synonym 2-nitro-p-toluidine, 4-amino-3-nitrotoluene, 3-nitro-4-toluidine, fast red gl, 3-nitro-4-aminotoluene, fast red gl base, benzenamine, 4-methyl-2-nitro, 2-nitro-4-methylaniline, 4-methyl-6-nitroaniline, mnpt
PubChem CID 6978
ChEBI CHEBI:66920
IUPAC Name 4-methyl-2-nitroaniline
SMILES CC1=CC(=C(C=C1)N)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N0114500G View Documents TCI America
N0114500G
500g Each for $314.73
 
N011425G View Documents TCI America
N011425G
25g Each for $42.78
 
Specifications

Chemical Identifiers

89-62-3
152.153
DLURHXYXQYMPLT-UHFFFAOYSA-N
6978
4-methyl-2-nitroaniline
C7H8N2O2
MFCD00007907
2-nitro-p-toluidine, 4-amino-3-nitrotoluene, 3-nitro-4-toluidine, fast red gl, 3-nitro-4-aminotoluene, fast red gl base, benzenamine, 4-methyl-2-nitro, 2-nitro-4-methylaniline, 4-methyl-6-nitroaniline, mnpt
CHEBI:66920
CC1=CC(=C(C=C1)N)[N+](=O)[O-]

Specifications

Undesignated
500g
C7H8N2O2
2660
DLURHXYXQYMPLT-UHFFFAOYSA-N
4-methyl-2-nitroaniline
6978
152.15
≥98.0% (GC)
116°C
89-62-3
MFCD00007907
2-nitro-p-toluidine, 4-amino-3-nitrotoluene, 3-nitro-4-toluidine, fast red gl, 3-nitro-4-aminotoluene, fast red gl base, benzenamine, 4-methyl-2-nitro, 2-nitro-4-methylaniline, 4-methyl-6-nitroaniline, mnpt
CC1=CC(=C(C=C1)N)[N+](=O)[O-]
152.153
CHEBI:66920
Crystal-Powder at 20°C
4-Methyl-2-nitroaniline
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0401&(5)-2302

RTECSNumber : XU8227250

TSCA : Yes