missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-2-hexylamine Hydrochloride, TCI America™

$29.03 - $67.22

Chemical Identifiers

CAS 13803-74-2
Molecular Formula C7H18ClN
Molecular Weight (g/mol) 151.678
MDL Number MFCD09038354
InChI Key ZKKBPHUAHARETG-UHFFFAOYSA-N
Synonym Methylhexanamine Hydrochloride, 1,3-Dimethylpentylamine Hydrochloride, 1,3-Dimethylamylamine Hydrochloride, 2-Amino-4-methylhexane Hydrochloride
PubChem CID 44118858
IUPAC Name 4-methylhexan-2-amine;hydrochloride
SMILES CCC(C)CC(C)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M24095G View Documents TCI America
M24095G
5g Each for $67.22
 
M24091G View Documents TCI America
M24091G
1g Each for $29.03
 
Specifications

Chemical Identifiers

13803-74-2
151.678
ZKKBPHUAHARETG-UHFFFAOYSA-N
44118858
CCC(C)CC(C)N.Cl
C7H18ClN
MFCD09038354
Methylhexanamine Hydrochloride, 1,3-Dimethylpentylamine Hydrochloride, 1,3-Dimethylamylamine Hydrochloride, 2-Amino-4-methylhexane Hydrochloride
4-methylhexan-2-amine;hydrochloride

Specifications

White-Yellow
5g
C7H18ClN
Methylhexanamine Hydrochloride, 1,3-Dimethylpentylamine Hydrochloride, 1,3-Dimethylamylamine Hydrochloride, 2-Amino-4-methylhexane Hydrochloride
CCC(C)CC(C)N.Cl
151.678
151.68
≥98.0% (T)
130°C
13803-74-2
MFCD09038354
ZKKBPHUAHARETG-UHFFFAOYSA-N
4-methylhexan-2-amine;hydrochloride
44118858
Crystal-Powder at 20°C
4-Methyl-2-hexylamine Hydrochloride
Safety and Handling

Safety and Handling

TSCA : No