missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Methyl-2-cyanobiphenyl, 98%, ACROS Organics™

Chemical Identifiers

CAS 114772-53-1
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD00151805
InChI Key ZGQVZLSNEBEHFN-UHFFFAOYSA-N
Synonym 2'-cyano-4-methylbiphenyl
PubChem CID 145512
IUPAC Name 2-(4-methylphenyl)benzonitrile
SMILES CC1=CC=C(C=C1)C2=CC=CC=C2C#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC358200050 SDS View Documents Thermo Fisher Scientific
AC358200050
5g Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

114772-53-1
193.249
ZGQVZLSNEBEHFN-UHFFFAOYSA-N
145512
CC1=CC=C(C=C1)C2=CC=CC=C2C#N
C14H11N
MFCD00151805
2'-cyano-4-methylbiphenyl
2-(4-methylphenyl)benzonitrile

Specifications

Authentic
White to Beige
5g
97.5
98%
MFCD00151805
ZGQVZLSNEBEHFN-UHFFFAOYSA-N
2-(4-methylphenyl)benzonitrile
145512
Crystalline Powder
4'-Methyl-2-cyanobiphenyl
Glass Bottle
49°C to 53°C
114772-53-1
100.0
C14H11N
2'-cyano-4-methylbiphenyl
CC1=CC=C(C=C1)C2=CC=CC=C2C#N
193.249
193.25
≥97.5%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning