missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-2,5-diphenylpyridine, TCI America™

$244.45 - $846.41

Chemical Identifiers

CAS 156021-08-8
Molecular Formula C18H15N
Molecular Weight (g/mol) 245.325
MDL Number MFCD13152024
InChI Key YHQCNNMCDMYPIA-UHFFFAOYSA-N
PubChem CID 14924986
IUPAC Name 4-methyl-2,5-diphenylpyridine
SMILES CC1=CC(=NC=C1C2=CC=CC=C2)C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M28265G View Documents TCI America
M28265G
5g Each for $846.41
 
M28261G View Documents TCI America
M28261G
1g Each for $244.45
 
Specifications

Chemical Identifiers

156021-08-8
245.325
YHQCNNMCDMYPIA-UHFFFAOYSA-N
4-methyl-2,5-diphenylpyridine
C18H15N
MFCD13152024
14924986
CC1=CC(=NC=C1C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Yellow
5g
C18H15N
YHQCNNMCDMYPIA-UHFFFAOYSA-N
4-methyl-2,5-diphenylpyridine
14924986
Crystal-Powder at 20°C
4-Methyl-2,5-diphenylpyridine
129°C
156021-08-8
MFCD13152024
CC1=CC(=NC=C1C2=CC=CC=C2)C3=CC=CC=C3
245.325
245.33
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No