Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-2,3,5,6-tetrafluorobenzyl bromide, 98%

Chemical Identifiers

Chemical Name or Material 4-Methyl-2,3,5,6-tetrafluorobenzyl bromide
CAS 92814-00-1
Molecular Formula C8H5BrF4
Formula Weight 257.03
MDL Number MFCD03001155
UN Number UN3261
Beilstein 5003900
Synonym 2,3,5,6-Tetrafluoro-4-methylbenzyl bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2021003

  SDS
alfa aesar™
L2021003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2021014

  SDS
alfa aesar™
L2021014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 257.03
Chemical Name or Material 4-Methyl-2,3,5,6-tetrafluorobenzyl bromide
Molecular Formula C8H5BrF4
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD03001155
Assay 98%
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Health Hazard 1 Danger
Boiling Point 49°C (0.4mmHg)
Synonym 2,3,5,6-Tetrafluoro-4-methylbenzyl bromide
Quantity 1g
UN Number UN3261
Beilstein 5003900
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Melting Point 41° to 43°C
CAS 92814-00-1
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.