Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-2-(2-thiazolylazo)phenol, 97%

Chemical Identifiers

Chemical Name or Material 4-Methyl-2-(2-thiazolylazo)phenol
CAS 1823-44-5
Formula Weight 219.26g/mol
MDL Number MFCD00005323
Beilstein 190372
Synonym 2-(2-Thiazolylazo)-p-cresol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5554403

 
alfa aesar™
H55544-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5554406

 
alfa aesar™
H55544-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5554414

 
alfa aesar™
H55544-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Orange to brown
Melting Point 130 to 132°C
Quantity 1g
Chemical Name or Material 4-Methyl-2-(2-thiazolylazo)phenol
CAS 1823-44-5
Physical Form Solid
Assay 97%
Formula Weight 219.26g/mol
MDL Number MFCD00005323
Beilstein 190372
Synonym 2-(2-Thiazolylazo)-p-cresol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for sev
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for sev

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.