Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-2-(2-pyridyl)thiazole-5-carboxylic acid, 97%

$83.77 - $342.04

Chemical Identifiers

Chemical Name or Material 4-Methyl-2-(2-pyridyl)thiazole-5-carboxylic acid
CAS 34418-48-9
Molecular Formula C10H8N2O2S
Formula Weight 220.25
MDL Number MFCD01316364
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5171103

  SDS
alfa aesar™
H5171103
1g Each for $83.77

AAH5171106

  SDS
alfa aesar™
H5171106
5g Each for $342.04
Description & Specifications

Specifications

Formula Weight 220.25
Chemical Name or Material 4-Methyl-2-(2-pyridyl)thiazole-5-carboxylic acid
Assay 97%
Molecular Formula C10H8N2O2S
MDL Number MFCD01316364
CAS 34418-48-9
Health Hazard 1 Warning
Melting Point 224° to 228°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.