missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methyl-2,1,3-benzoxadiazole, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC441360010
Specifications
4-Methyl-2, 1, 3-benzoxadiazole | |
Glass Bottle | |
40.0°C to 41.0°C | |
29091-40-5 | |
100.0 | |
C7H6N2O | |
PTXJEYIAUIOTSH-UHFFFAOYSA-N | |
4-methyl-2,1,3-benzoxadiazole | |
584114 | |
Crystalline Powder or Lms |
Authentic | |
Yellow | |
1g | |
94.0 | |
96% min. (GC) | |
#, 2,1,3-benzoxadiazole, 4-methyl, 2,1,3-benzoxadiazole,4-methyl, 4-methyl-benzo 1,2,5 oxadiazole, 4-methyl-benzofurazan, 4-methylbenzo c 1,2,5 oxadiazole, 4-methylbenzofurazan, methylbenzfurazan | |
CC1=CC=CC2=NON=C12 | |
134.14 | |
134.14 | |
95% |
Chemical Identifiers
29091-40-5 | |
134.14 | |
#, 2,1,3-benzoxadiazole, 4-methyl, 2,1,3-benzoxadiazole,4-methyl, 4-methyl-benzo 1,2,5 oxadiazole, 4-methyl-benzofurazan, 4-methylbenzo c 1,2,5 oxadiazole, 4-methylbenzofurazan, methylbenzfurazan | |
4-methyl-2,1,3-benzoxadiazole |
C7H6N2O | |
PTXJEYIAUIOTSH-UHFFFAOYSA-N | |
584114 | |
CC1=CC=CC2=NON=C12 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
GHS Signal Word: Warning