Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-1-tetralone, 97%

$56.64 - $478.72

Chemical Identifiers

Chemical Name or Material 4-Methyl-1-tetralone
CAS 19832-98-5
Molecular Formula C11H12O
Formula Weight 160.22
MDL Number MFCD00001691
Synonym 4-Methyl-1,2,3,4-tetrahydro-1-naphthalenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2196703

  SDS
alfa aesar™
B2196703
1g Each for $56.64

AAB2196706

  SDS
alfa aesar™
B2196706
5g Each for $130.22

AAB2196714

  SDS
alfa aesar™
B2196714
25g Each for $478.72
Description & Specifications

Specifications

Formula Weight 160.22
Chemical Name or Material 4-Methyl-1-tetralone
Assay 97%
Molecular Formula C11H12O
MDL Number MFCD00001691
CAS 19832-98-5
Synonym 4-Methyl-1,2,3,4-tetrahydro-1-naphthalenone
Flash Point >110°C (230°F)
Boiling Point 95°C (1mmHg)
Quantity 1g

Applications
4-Methyl-1-tetralone is an important intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.