missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-1-piperazineacetonitrile, 97%

Manufacturer:  Alfa Aesar™ H6394903

 View more versions of this product

Catalog No. AAH6394903



Specifications

Specifications

4-Methyl-1-piperazineacetonitrile
874-77-1
UN3439
XCUQQNRWWCWVOP-UHFFFAOYSA-N
2-(4-methylpiperazin-1-yl)acetonitrile
10582860
Solid
1g
C7H13N3
2-4-methylpiperazin-1-yl acetonitrile, 4-methylpiperazin-1-yl acetonitrile, 4-methyl-1-piperazineacetonitrile, 1-4-methylpiperazine acetonitrile, 1-cyanomethyl-4-methylpiperazine, 1-piperazineacetonitrile, 4-methyl, 4-methyl-piperazin-yl-acetonitrile, 4-methyl-piperazin-1-yl-acetonitrile
CN1CCN(CC1)CC#N
139.202
139.2
97%

Chemical Identifiers

874-77-1
139.202
2-4-methylpiperazin-1-yl acetonitrile, 4-methylpiperazin-1-yl acetonitrile, 4-methyl-1-piperazineacetonitrile, 1-4-methylpiperazine acetonitrile, 1-cyanomethyl-4-methylpiperazine, 1-piperazineacetonitrile, 4-methyl, 4-methyl-piperazin-yl-acetonitrile, 4-methyl-piperazin-1-yl-acetonitrile
2-(4-methylpiperazin-1-yl)acetonitrile
C7H13N3
XCUQQNRWWCWVOP-UHFFFAOYSA-N
10582860
CN1CCN(CC1)CC#N
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

SDS Documents