Learn More
4-Methyl-1-piperazineacetonitrile, 97%, Thermo Scientific™
$72.33 - $207.99
Chemical Identifiers
CAS | 874-77-1 |
---|---|
Molecular Formula | C7H13N3 |
Molecular Weight (g/mol) | 139.202 |
InChI Key | XCUQQNRWWCWVOP-UHFFFAOYSA-N |
Synonym | 2-4-methylpiperazin-1-yl acetonitrile, 4-methylpiperazin-1-yl acetonitrile, 4-methyl-1-piperazineacetonitrile, 1-4-methylpiperazine acetonitrile, 1-cyanomethyl-4-methylpiperazine, 1-piperazineacetonitrile, 4-methyl, 4-methyl-piperazin-yl-acetonitrile, 4-methyl-piperazin-1-yl-acetonitrile |
PubChem CID | 10582860 |
IUPAC Name | 2-(4-methylpiperazin-1-yl)acetonitrile |
SMILES | CN1CCN(CC1)CC#N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH63949MD
|
Thermo Scientific Chemicals
H63949MD |
250 mg |
Each for $72.33
|
|
|||||
AAH6394903
|
Thermo Scientific Chemicals
H6394903 |
1 g |
Each for $207.99
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
874-77-1 | |
139.202 | |
2-4-methylpiperazin-1-yl acetonitrile, 4-methylpiperazin-1-yl acetonitrile, 4-methyl-1-piperazineacetonitrile, 1-4-methylpiperazine acetonitrile, 1-cyanomethyl-4-methylpiperazine, 1-piperazineacetonitrile, 4-methyl, 4-methyl-piperazin-yl-acetonitrile, 4-methyl-piperazin-1-yl-acetonitrile | |
2-(4-methylpiperazin-1-yl)acetonitrile |
C7H13N3 | |
XCUQQNRWWCWVOP-UHFFFAOYSA-N | |
10582860 | |
CN1CCN(CC1)CC#N |
Specifications
874-77-1 | |
250 mg | |
2-4-methylpiperazin-1-yl acetonitrile, 4-methylpiperazin-1-yl acetonitrile, 4-methyl-1-piperazineacetonitrile, 1-4-methylpiperazine acetonitrile, 1-cyanomethyl-4-methylpiperazine, 1-piperazineacetonitrile, 4-methyl, 4-methyl-piperazin-yl-acetonitrile, 4-methyl-piperazin-1-yl-acetonitrile | |
CN1CCN(CC1)CC#N | |
139.202 | |
139.2 | |
Solid |
C7H13N3 | |
UN3439 | |
XCUQQNRWWCWVOP-UHFFFAOYSA-N | |
2-(4-methylpiperazin-1-yl)acetonitrile | |
10582860 | |
97% | |
4-Methyl-1-piperazineacetonitrile |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures