missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-1-phenyl-2-pentanol, TCI America™

$58.06 - $163.47

Chemical Identifiers

CAS 7779-78-4
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.275
MDL Number MFCD00209515
InChI Key IUADYGVMSDKSMB-UHFFFAOYSA-N
PubChem CID 62661
IUPAC Name 4-methyl-1-phenylpentan-2-ol
SMILES CC(C)CC(CC1=CC=CC=C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M201225G View Documents TCI America
M201225G
25g Each for $163.47
 
M20125G View Documents TCI America
M20125G
5g Each for $58.06
 
Specifications

Chemical Identifiers

7779-78-4
178.275
IUADYGVMSDKSMB-UHFFFAOYSA-N
4-methyl-1-phenylpentan-2-ol
C12H18O
MFCD00209515
62661
CC(C)CC(CC1=CC=CC=C1)O

Specifications

123°C
25g
C12H18O
IUADYGVMSDKSMB-UHFFFAOYSA-N
4-methyl-1-phenylpentan-2-ol
62661
Clear Liquid at 20°C
4-Methyl-1-phenyl-2-pentanol
Colorless
7779-78-4
MFCD00209515
CC(C)CC(CC1=CC=CC=C1)O
178.275
178.28
≥96.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1044

RTECSNumber : DA0513300

TSCA : Yes