Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-1-pentyn-3-ol, 97%

Chemical Identifiers

Chemical Name or Material 4-Methyl-1-pentyn-3-ol
CAS 565-68-4
Molecular Formula C6H10O
Formula Weight 98.15
MDL Number MFCD00039847
UN Number UN1987
Beilstein 1698762
Synonym Ethynyl isopropyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0531809

  SDS
alfa aesar™
L0531809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 98.15
Molecular Formula C6H10O
Chemical Name or Material 4-Methyl-1-pentyn-3-ol
Assay 97%
MDL Number MFCD00039847
CAS 565-68-4
UN Number UN1987
Synonym Ethynyl isopropyl carbinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P261
Avoid breathing dust/fume/gas/mist/vapors/spray.
Flash Point 41°C (105°F)
Boiling Point 133° to 134°C
Quantity 10g
Beilstein 1698762
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
Odor Recognizable

Applications
4-Methyl-1-pentyn-3-ol, is used as standard Pharmaceutical intermediate and also used in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261
Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS H Statement
H226
Flammable liquid and vapor.