Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-1-pentenylboronic acid, 97%

$144.80 - $144.80

Chemical Identifiers

Chemical Name or Material 4-Methyl-1-pentenylboronic acid
CAS 214907-33-2
Molecular Formula C6H13BO2
Formula Weight 127.98
MDL Number MFCD01074680
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27655MD

  SDS
alfa aesar™
H27655MD
250mg Each for $144.80
Description & Specifications

Specifications

Formula Weight 127.98
Molecular Formula C6H13BO2
MDL Number MFCD01074680
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Methyl-1-pentenylboronic acid
CAS 214907-33-2
Melting Point 99° to 102°C
Assay 97%
Health Hazard 1 Warning
Quantity 250mg
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.