Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-1-pentene, 97%

Chemical Identifiers

Chemical Name or Material 4-Methyl-1-pentene
CAS 691-37-2
Molecular Formula C6H12
Formula Weight 84.16
MDL Number MFCD00008949
UN Number UN3295
Beilstein 1731096
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0337214

  SDS
alfa aesar™
L0337214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 84.16
Health Hazard 3 GHS P Statement
P210-P243-P301+P330+P331-P315-P403+P233
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Take precautionary measures against static discharge.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Get immediate medical advice/attention.
Store in a well-ventilated place.
Keep container tightly closed.
Chemical Name or Material 4-Methyl-1-pentene
MDL Number MFCD00008949
Assay 97%
Health Hazard 1 Danger
CAS 691-37-2
Molecular Formula C6H12
Health Hazard 2 GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Melting Point −153°C
TSCA TSCA
Quantity 25g
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Beilstein 1731096
Boiling Point 53° to 54°C
Odor Petrol-like
Flash Point −32°C (−25°F)
UN Number UN3295
Safety and Handling

Danger

GHS P Statement
P210-P243-P301+P330+P331-P315-P403+P233
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Take precautionary measures against static discharge.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Get immediate medical advice/attention.
Store in a well-ventilated place.
Keep container tightly closed.

GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.