missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-1-naphthoyl chloride, 98%

$28.84 - $422.54

Chemical Identifiers

CAS 87700-67-2
Molecular Formula C12H9ClO
Molecular Weight (g/mol) 204.653
MDL Number MFCD12025362
InChI Key IXGMHYHWSPDNNS-UHFFFAOYSA-N
Synonym 1-naphthalenecarbonylchloride, 4-methyl, 1-naphthalenecarbonyl chloride, 4-methyl, 4-methyl-1-naphthalenecarbonyl chloride, 1-naphthalenecarbonyl chloride, 4-methyl-9ci, 4-methyl-1-napthoyl chloride, 4-methyl-1-naphthoyl chloride, 4-methyl-naphthalene-1-carbonyl chloride
PubChem CID 24887412
IUPAC Name 4-methylnaphthalene-1-carbonyl chloride
SMILES CC1=CC=C(C2=CC=CC=C12)C(=O)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6491703 View Documents Alfa Aesar™
H6491703
1g Each for $28.84
 
AAH6491706 View Documents Alfa Aesar™
H6491706
5g Each for $106.26
 
AAH6491714 View Documents Alfa Aesar™
H6491714
25g Each for $422.54
 
Specifications

Chemical Identifiers

87700-67-2
204.653
IXGMHYHWSPDNNS-UHFFFAOYSA-N
24887412
CC1=CC=C(C2=CC=CC=C12)C(=O)Cl
C12H9ClO
MFCD12025362
1-naphthalenecarbonylchloride, 4-methyl, 1-naphthalenecarbonyl chloride, 4-methyl, 4-methyl-1-naphthalenecarbonyl chloride, 1-naphthalenecarbonyl chloride, 4-methyl-9ci, 4-methyl-1-napthoyl chloride, 4-methyl-1-naphthoyl chloride, 4-methyl-naphthalene-1-carbonyl chloride
4-methylnaphthalene-1-carbonyl chloride

Specifications

Yellow
87700-67-2
MFCD12025362
3247818
IXGMHYHWSPDNNS-UHFFFAOYSA-N
4-methylnaphthalene-1-carbonyl chloride
24887412
Solid
4-Methyl-1-naphthoyl chloride
1g
C12H9ClO
UN3261
1-naphthalenecarbonylchloride, 4-methyl, 1-naphthalenecarbonyl chloride, 4-methyl, 4-methyl-1-naphthalenecarbonyl chloride, 1-naphthalenecarbonyl chloride, 4-methyl-9ci, 4-methyl-1-napthoyl chloride, 4-methyl-1-naphthoyl chloride, 4-methyl-naphthalene-1-carbonyl chloride
CC1=CC=C(C2=CC=CC=C12)C(=O)Cl
204.653
204.65
98%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.