missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-1-naphthoic acid, 97%

$31.93 - $260.62

Chemical Identifiers

CAS 4488-40-8
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00671557
InChI Key SIVYRLBDAPKADZ-UHFFFAOYSA-N
Synonym 4-methyl-1-naphthoic acid, 4-methyl-1-naphthalenecarboxylic acid, 4-methyl-1-napthoic acid, 1-naphthalenecarboxylic acid, 4-methyl, 4-methyl-naphthalene-1-carboxylic acid, 4-methylnaphthalenecarboxylic acid, 4-methylnaphthoic acid, acmc-1aehw, 4-methyl-1-naphthoicacid, 4-methyl-1-naphtholic acid
PubChem CID 3014285
IUPAC Name 4-methylnaphthalene-1-carboxylic acid
SMILES CC1=CC=C(C2=CC=CC=C12)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6076502 View Documents Alfa Aesar™
H6076502
0.5g Each for $31.93
 
AAH6076509 View Documents Alfa Aesar™
H6076509
10g Each for $260.62
 
Specifications

Chemical Identifiers

4488-40-8
186.21
SIVYRLBDAPKADZ-UHFFFAOYSA-N
3014285
CC1=CC=C(C2=CC=CC=C12)C(=O)O
C12H10O2
MFCD00671557
4-methyl-1-naphthoic acid, 4-methyl-1-naphthalenecarboxylic acid, 4-methyl-1-napthoic acid, 1-naphthalenecarboxylic acid, 4-methyl, 4-methyl-naphthalene-1-carboxylic acid, 4-methylnaphthalenecarboxylic acid, 4-methylnaphthoic acid, acmc-1aehw, 4-methyl-1-naphthoicacid, 4-methyl-1-naphtholic acid
4-methylnaphthalene-1-carboxylic acid

Specifications

179°C to 181°C
4488-40-8
MFCD00671557
SIVYRLBDAPKADZ-UHFFFAOYSA-N
4-methylnaphthalene-1-carboxylic acid
3014285
97%
0.5g
C12H10O2
4-methyl-1-naphthoic acid, 4-methyl-1-naphthalenecarboxylic acid, 4-methyl-1-napthoic acid, 1-naphthalenecarboxylic acid, 4-methyl, 4-methyl-naphthalene-1-carboxylic acid, 4-methylnaphthalenecarboxylic acid, 4-methylnaphthoic acid, acmc-1aehw, 4-methyl-1-naphthoicacid, 4-methyl-1-naphtholic acid
CC1=CC=C(C2=CC=CC=C12)C(=O)O
186.21
186.21
4-Methyl-1-naphthoic acid