missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-1-naphthaleneboronic Acid (contains varying amounts of Anhydride), TCI America™
$76.43 - $76.43
Chemical Identifiers
| CAS | 103986-53-4 |
|---|---|
| Molecular Formula | C11H11BO2 |
| Molecular Weight (g/mol) | 186.02 |
| MDL Number | MFCD01632204 |
| InChI Key | JHVQEUGNYSVSDH-UHFFFAOYSA-N |
| PubChem CID | 2773511 |
| IUPAC Name | (4-methylnaphthalen-1-yl)boronic acid |
| SMILES | CC1=CC=C(B(O)O)C2=CC=CC=C12 |
Chemical Identifiers
| 103986-53-4 | |
| 186.02 | |
| JHVQEUGNYSVSDH-UHFFFAOYSA-N | |
| (4-methylnaphthalen-1-yl)boronic acid |
| C11H11BO2 | |
| MFCD01632204 | |
| 2773511 | |
| CC1=CC=C(B(O)O)C2=CC=CC=C12 |
Specifications
| 103986-53-4 | |
| Yellow | |
| MFCD01632204 | |
| 4-methyl-1-naphthaleneboronic acid, 4-methyl-1-naphthalene boronic acid, 4-methylnaphthalene-1-boronic acid, 1-methylnaphthalene-4-boronic acid, 4-methylnaphthalen-1-yl boronic acid, 1-borono-4-methylnaphthalene, 4-methyl-1-naphthyl boronic acid, 4-methyl-1-naphthylboronic acid, 4-methylnaphthalen-1-yl boranediol, boronic acid, 4-methyl-1-naphthalenyl | |
| CC1=CC=C(B(O)O)C2=CC=CC=C12 | |
| 186.02 | |
| 186.02 | |
| 4-Methyl-1-naphthaleneboronic Acid (contains varying amounts of Anhydride) |
| 207°C | |
| C11H11BO2 | |
| 1 g | |
| JHVQEUGNYSVSDH-UHFFFAOYSA-N | |
| (4-methylnaphthalen-1-yl)boronic acid | |
| 2773511 | |
| Crystalline Powder |
Safety and Handling
TSCA : No