missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-1-naphthaleneboronic acid, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC437530010

 View more versions of this product

Catalog No. AC437530010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Methyl-1-naphthaleneboronic acid
Glass Bottle
224°C to 228°C
103986-53-4
100.0
C11H11BO2
JHVQEUGNYSVSDH-UHFFFAOYSA-N
(4-methylnaphthalen-1-yl)boronic acid
2773511
Powder
Authentic
White
1g
95.0
95% min. (HPLC)
1-borono-4-methylnaphthalene, 1-methylnaphthalene-4-boronic acid, 4-methyl-1-naphthalene boronic acid, 4-methyl-1-naphthaleneboronic acid, 4-methyl-1-naphthyl boronic acid, 4-methyl-1-naphthylboronic acid, 4-methylnaphthalen-1-yl boranediol, 4-methylnaphthalen-1-yl boronic acid, 4-methylnaphthalene-1-boronic acid, boronic acid, 4-methyl-1-naphthalenyl
B(C1=CC=C(C2=CC=CC=C12)C)(O)O
186.02
186.02
96%

Chemical Identifiers

103986-53-4
186.02
1-borono-4-methylnaphthalene, 1-methylnaphthalene-4-boronic acid, 4-methyl-1-naphthalene boronic acid, 4-methyl-1-naphthaleneboronic acid, 4-methyl-1-naphthyl boronic acid, 4-methyl-1-naphthylboronic acid, 4-methylnaphthalen-1-yl boranediol, 4-methylnaphthalen-1-yl boronic acid, 4-methylnaphthalene-1-boronic acid, boronic acid, 4-methyl-1-naphthalenyl
(4-methylnaphthalen-1-yl)boronic acid
C11H11BO2
JHVQEUGNYSVSDH-UHFFFAOYSA-N
2773511
B(C1=CC=C(C2=CC=CC=C12)C)(O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents