missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-1-indanone, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC439410010

 View more versions of this product

Catalog No. AC439410010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Methyl-1-indanone
White to Yellow
1g
96.0
97%
4-methyl-1-indanone, 4-methyl-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 2,3-dihydro-4-methyl, 4-methylindan-1-one, 4-methyl-indanone, 4-methylindanone, methyl-1-indanone, 4-methylindan-1-on, pubchem8834, 4-methyl-indan-1-one
CC1=CC=CC2=C1CCC2=O
146.189
146.19
≥96.0%
Authentic
94°C to 101°C
24644-78-8
100.0
C10H10O
RUORWXQKVXTQJJ-UHFFFAOYSA-N
4-methyl-2,3-dihydroinden-1-one
4418431
Crystalline Powder

Chemical Identifiers

24644-78-8
146.189
4-methyl-1-indanone, 4-methyl-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 2,3-dihydro-4-methyl, 4-methylindan-1-one, 4-methyl-indanone, 4-methylindanone, methyl-1-indanone, 4-methylindan-1-on, pubchem8834, 4-methyl-indan-1-one
4-methyl-2,3-dihydroinden-1-one
C10H10O
RUORWXQKVXTQJJ-UHFFFAOYSA-N
4418431
CC1=CC=CC2=C1CCC2=O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents